siRNA Details
| Gene Symbol: Hoxa4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008265.3 | NP_032291.1 | 2 | 795 | ||
| GenBank mRNA | AK076098.1 | - | 2 | 796 | ||
| BC107172.2 | - | 3 | 502 | 40044111 | ||
| M13813.1 | - | 1 | 240 | |||
| M27432.1 | - | 1 | 243 | |||
| X13538.1 | - | 1 | 243 | |||

Hoxa4
Genomic Map