siRNA Details
Gene Symbol: Hoxa5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010453.5 | NP_034583.1 | 1 | 326 | ||
GenBank mRNA | AB221550.1 | - | 1 | 89 | ||
AK144356.1 | - | 1 | 67 | |||
BC011063.1 | - | 1 | 150 | 3985274 | ||
M28021.1 | - | 1 | 326 | |||
M36604.1 | - | 1 | 203 | |||
X16840.1 | - | 1 | 203 |
