siRNA Details
| Gene Symbol: Hoxb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079869.1 | NP_001073338.1 | 4 | 1610 | ||
| NM_010458.2 | NP_034588.2 | 2 | 1533 | |||
| GenBank mRNA | S35628.1 | - | 2 | 1533 | ||
| S35738.1 | - | 4 | 1611 | |||
| X66177.1 | - | 2 | 1533 | |||

Hoxb3
Genomic Map