siRNA Details
| Gene Symbol: Hoxb7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010460.2 | NP_034590.2 | 1 | 174 | ||
| GenBank mRNA | AK136706.1 | - | 1 | 174 | ||
| X06762.1 | - | 2256 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hoxb7| Gene Symbol: Hoxb7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010460.2 | NP_034590.2 | 1 | 174 | ||
| GenBank mRNA | AK136706.1 | - | 1 | 174 | ||
| X06762.1 | - | 2256 | ||||