siRNA Details
| Gene Symbol: Hoxc8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010466.2 | NP_034596.1 | 1 | 406 | ||
| GenBank mRNA | AB221617.1 | - | 1 | 169 | ||
| X07439.1 | - | 1 | 406 | |||
| X07646.1 | - | 1 | 634 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hoxc8| Gene Symbol: Hoxc8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010466.2 | NP_034596.1 | 1 | 406 | ||
| GenBank mRNA | AB221617.1 | - | 1 | 169 | ||
| X07439.1 | - | 1 | 406 | |||
| X07646.1 | - | 1 | 634 | |||