siRNA Details
| Gene Symbol: Hoxd12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008274.3 | NP_032300.2 | 1 | 453 | ||
| GenBank mRNA | AK031206.1 | - | 1 | 392 | ||
| BC145656.1 | - | 1 | 453 | 40130959 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hoxd12| Gene Symbol: Hoxd12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008274.3 | NP_032300.2 | 1 | 453 | ||
| GenBank mRNA | AK031206.1 | - | 1 | 392 | ||
| BC145656.1 | - | 1 | 453 | 40130959 | ||