siRNA Details
| Gene Symbol: Hoxd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010468.2 | NP_034598.2 | 2 | 537 | ||
| GenBank mRNA | AK133705.1 | - | 2 | 405 | ||
| AK138525.1 | - | 2 | 538 | |||
| BC137716.1 | - | 1 | 162 | 8860735 | ||
| BC137718.1 | - | 1 | 162 | 8860737 | ||
| U56400.1 | - | 1 | 213 | |||
| X73573.1 | - | 1 | 201 | |||

Hoxd3
Genomic Map