siRNA Details
Gene Symbol: Eif2ak1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013557.2 | NP_038585.2 | 508 | |||
GenBank mRNA | AK014775.1 | - | 470 | |||
AK155206.1 | - | 490 | ||||
AK156492.1 | - | 469 | ||||
AK158437.1 | - | 464 | ||||
AK173160.1 | - | 458 | ||||
AY033898.1 | - | 493 | ||||
BB625879.1 | - | 509 | ||||
BC028923.1 | - | 480 | 4164417 | |||
BC111035.1 | - | 459 | 5357121 |
