siRNA Details
Gene Symbol: Dnajb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008299.3 | NP_032325.2 | 1 | 548 | ||
GenBank mRNA | AK005688.1 | - | 1 | 506 | ||
AK005690.1 | - | 1 | 497 | |||
BC048490.1 | - | 1 | 516 | 6775233 | ||
U95607.1 | - | 1 | 550 |
