siRNA Details
| Gene Symbol: Hspg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008305.3 | NP_032331.2 | 33 | 4207 | ||
| GenBank mRNA | AK147294.1 | - | 33 | 4208 | ||
| AK147396.1 | - | 33 | 4208 | |||
| J04054.1 | - | 11 | 1331 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hspg2| Gene Symbol: Hspg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008305.3 | NP_032331.2 | 33 | 4207 | ||
| GenBank mRNA | AK147294.1 | - | 33 | 4208 | ||
| AK147396.1 | - | 33 | 4208 | |||
| J04054.1 | - | 11 | 1331 | |||