siRNA Details
| Gene Symbol: Htr1f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008310.3 | NP_032336.1 | 3 | 657 | ||
| GenBank mRNA | AK045364.1 | - | 3 | 726 | ||
| BC120507.1 | - | 3 | 513 | 40129430 | ||
| BC137604.1 | - | 3 | 512 | 8860623 | ||
| Z14224.1 | - | 3 | 656 | |||
More Information
Set Membership: |
|

Htr1f
Genomic Map