siRNA Details
| Gene Symbol: Hus1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF076845.1 | - | 2 | 134 | ||
| AK046152.1 | - | 2 | 144 | |||
| AK051839.1 | - | 1 | 569 | |||
| AK135922.1 | - | 2 | 124 | |||
| BB611267.1 | - | 2 | 200 | |||
| BC047460.1 | - | 2 | 151 | |||
| Y16894.2 | - | 2 | 166 | |||

Hus1
Genomic Map