siRNA Details
| Gene Symbol: Hus1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF076845.1 | - | 7, 8 | 773 | ||
| AK046152.1 | - | 7, 8 | 780 | |||
| AK051839.1 | - | 6, 7 | 1205 | |||
| AK088403.1 | - | 7, 8 | 783 | |||
| AK135922.1 | - | 7, 8 | 763 | |||
| BC047460.1 | - | 7, 8 | 790 | |||
| Y16894.2 | - | 7, 8 | 805 | |||

Hus1
Genomic Map