siRNA Details
| Gene Symbol: Icam2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010494.1 | NP_034624.1 | 2, 3 | 485 | ||
| GenBank mRNA | AK007801.1 | - | 2, 3 | 420 | ||
| BC039128.1 | - | 2, 3 | 417 | 3962648 | ||
| BC039970.1 | - | 2, 3 | 437 | 4487248 | ||
| X65493.1 | - | 2, 3 | 485 | |||

Icam2
Genomic Map