siRNA Details
| Gene Symbol: Id2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010496.3 | NP_034626.1 | 1 | 181 | ||
| GenBank mRNA | AK003222.1 | - | 1 | 178 | ||
| AK013239.1 | - | 1 | 176 | |||
| AK013378.1 | - | 1 | 179 | |||
| AK135547.1 | - | 1 | 183 | |||
| AK146346.1 | - | 1 | 181 | |||
| AK146606.1 | - | 1 | 184 | |||
| BC006921.1 | - | 1 | 158 | 2655173 | ||
| BC053699.1 | - | 1 | 168 | 6515664 | ||
| M69293.1 | - | 1 | 175 | |||

Id2
Genomic Map