siRNA Details
| Gene Symbol: Id2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010496.3 | NP_034626.1 | 1, 2 | 425 | ||
| GenBank mRNA | AK003222.1 | - | 1, 2 | 422 | ||
| AK013378.1 | - | 1, 2 | 422 | |||
| AK135547.1 | - | 1, 2 | 427 | |||
| AK146346.1 | - | 1, 2 | 425 | |||
| AK146606.1 | - | 1, 2 | 428 | |||
| BC006921.1 | - | 1, 2 | 402 | 2655173 | ||
| BC053699.1 | - | 1, 2 | 412 | 6515664 | ||
| M69293.1 | - | 1, 2 | 419 | |||

Id2
Genomic Map