siRNA Details
Gene Symbol: Ier5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010500.2 | NP_034630.1 | 1 | 949 | ||
GenBank mRNA | AF079527.1 | - | 1 | 949 | ||
AK146609.1 | - | 1 | 946 | |||
AK150461.1 | - | 1 | 948 | |||
BC051986.1 | - | 1 | 923 | 5707524 |
