siRNA Details
| Gene Symbol: Ifrd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025903.2 | NP_080179.1 | 10 | 1224 | ||
| GenBank mRNA | AK002702.1 | - | 4 | 449 | ||
| AK007687.1 | - | 10 | 1225 | |||
| AK149496.1 | - | 10 | 1205 | |||
| AK156625.1 | - | 9 | 1300 | |||
| AK165967.1 | - | 10 | 1218 | |||
| AK166130.1 | - | 10 | 1292 | |||
| AK166258.1 | - | 10 | 1182 | |||
| AY298860.1 | - | 10 | 1053 | |||
| BC023486.1 | - | 10 | 1171 | |||
| BC037434.1 | - | 10 | 1180 | 2651172 | ||

Ifrd2
Genomic Map