siRNA Details
| Gene Symbol: Ifrd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025903.2 | NP_080179.1 | 5, 6 | 718 | ||
| GenBank mRNA | AK007687.1 | - | 5, 6 | 719 | ||
| AK156625.1 | - | 4, 5 | 794 | |||
| AK165967.1 | - | 5, 6 | 712 | |||
| AK166130.1 | - | 5, 6 | 786 | |||
| AK166258.1 | - | 5, 6 | 676 | |||
| AK178654.1 | - | 2, 3 | 189 | |||
| AY298860.1 | - | 5, 6 | 547 | |||
| BC023486.1 | - | 5, 6 | 665 | |||
| BC037434.1 | - | 5, 6 | 674 | 2651172 | ||

Ifrd2
Genomic Map