siRNA Details
| Gene Symbol: Igfbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008341.4 | NP_032367.3 | 3, 4 | 863 | ||
| GenBank mRNA | BC013345.1 | - | 3, 4 | 894 | 4161889 | |
| CF616646.1 | - | 70 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Igfbp1| Gene Symbol: Igfbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008341.4 | NP_032367.3 | 3, 4 | 863 | ||
| GenBank mRNA | BC013345.1 | - | 3, 4 | 894 | 4161889 | |
| CF616646.1 | - | 70 | ||||