siRNA Details
| Gene Symbol: Jchain | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152839.3 | NP_690052.2 | 2 | 268 | ||
| GenBank mRNA | AB644392.1 | - | 2 | 150 | ||
| AK078865.1 | - | 2 | 154 | |||
| AK160344.1 | - | 2 | 220 | |||
| BC006026.1 | - | 2 | 172 | 3491646 | ||
| J00544.1 | - | 2 | 155 | |||
| M90766.1 | - | 2 | 171 | |||

Jchain
Genomic Map