siRNA Details
| Gene Symbol: Il10rb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008349.5 | NP_032375.2 | 5 | 650 | ||
| GenBank mRNA | AF440787.1 | - | 5 | 577 | ||
| AK040938.1 | - | 5 | 566 | |||
| AK134042.1 | - | 5 | 643 | |||
| AK160991.1 | - | 5 | 1605 | |||
| AK181086.1 | - | 1 | 27 | |||
| AK183290.1 | - | 1 | 27 | |||
| AK196209.1 | - | 2 | 137 | |||
| AK201410.1 | - | 1 | 27 | |||
| BC145791.1 | - | 5 | 650 | 40131094 | ||
| U53696.1 | - | 5 | 563 | |||

Il10rb
Genomic Map