siRNA Details
| Gene Symbol: Il18rap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010553.4 | NP_034683.1 | 7 | 1043 | ||
| GenBank mRNA | AF077347.1 | - | 7 | 1052 | ||
| AK137541.1 | - | 7 | 1004 | |||
| BC120598.1 | - | 7 | 1050 | 40129521 | ||
| BC120600.1 | - | 7 | 1050 | 40129523 | ||

Il18rap
Genomic Map