siRNA Details
| Gene Symbol: Il1r1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001123382.1 | NP_001116854.1 | 5 | 934 | ||
| NM_008362.2 | NP_032388.1 | 5 | 865 | |||
| GenBank mRNA | AK048550.1 | - | 5 | 852 | ||
| AK054137.1 | - | 5 | 935 | |||
| BC109135.2 | - | 5 | 771 | 40039321 | ||
| M20658.1 | - | 5 | 865 | |||

Il1r1
Genomic Map