siRNA Details
| Gene Symbol: Insr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010568.2 | NP_034698.2 | 11 | 2784 | ||
| GenBank mRNA | EU346716.1 | - | 5 | 636 | ||
| J05149.1 | - | 11 | 2343 | |||
| KC356871.1 | - | 12 | 2331 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Insr| Gene Symbol: Insr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010568.2 | NP_034698.2 | 11 | 2784 | ||
| GenBank mRNA | EU346716.1 | - | 5 | 636 | ||
| J05149.1 | - | 11 | 2343 | |||
| KC356871.1 | - | 12 | 2331 | |||