siRNA Details
| Gene Symbol: Eif3e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008388.2 | NP_032414.1 | 8, 9 | 894 | ||
| GenBank mRNA | AK002576.1 | - | 8, 9 | 871 | ||
| AK011436.1 | - | 8, 9 | 864 | |||
| AK088104.1 | - | 8, 9 | 871 | |||
| AK088269.1 | - | 8, 9 | 868 | |||
| AK145059.1 | - | 8, 9 | 877 | |||
| AK146936.1 | - | 8, 9 | 866 | |||
| AK161650.1 | - | 8, 9 | 864 | |||
| AK165929.1 | - | 8, 9 | 872 | |||
| AK181127.1 | - | 2, 3 | 248 | |||
| BC010252.1 | - | 8, 9 | 877 | |||
| BC029177.1 | - | 8, 9 | 855 | 3988386 | ||

Eif3e
Genomic Map