siRNA Details
| Gene Symbol: Invs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001281977.1 | NP_001268906.1 | 11 | 1692 | ||
| NM_001281978.1 | NP_001268907.1 | 13 | 1970 | |||
| NM_010569.4 | NP_034699.3 | 11 | 1744 | |||
| GenBank mRNA | AF034860.3 | - | 11 | 1686 | ||
| AJ010902.1 | - | 11 | 1685 | |||
| AK158766.1 | - | 13 | 1929 | |||
| BC126929.1 | - | 11 | 1581 | 40081431 | ||

Invs
Genomic Map