siRNA Details
| Gene Symbol: Itm2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008410.2 | NP_032436.1 | 3 | 522 | ||
| GenBank mRNA | AB030203.1 | - | 3 | 542 | ||
| AB030204.1 | - | 2 | 338 | |||
| AK002599.1 | - | 3 | 485 | |||
| AK003224.1 | - | 3 | 489 | |||
| AK003587.1 | - | 3 | 511 | |||
| AK003928.1 | - | 3 | 510 | |||
| AK005125.1 | - | 3 | 513 | |||
| AK076139.1 | - | 3 | 531 | |||
| AK077482.1 | - | 3 | 524 | |||
| AK150378.1 | - | 3 | 542 | |||
| AK150396.1 | - | 3 | 541 | |||
| AK151714.1 | - | 3 | 541 | |||
| AK151889.1 | - | 3 | 542 | |||
| AK152068.1 | - | 3 | 530 | |||
| AK152516.1 | - | 3 | 542 | |||
| AK152650.1 | - | 3 | 542 | |||
| AK152731.1 | - | 3 | 513 | |||
| AK152983.1 | - | 3 | 534 | |||
| AK153103.1 | - | 3 | 485 | |||
| AK159307.1 | - | 3 | 485 | |||
| AK159340.1 | - | 3 | 510 | |||
| AK159378.1 | - | 3 | 541 | |||
| AK159426.1 | - | 3 | 510 | |||
| AK159619.1 | - | 3 | 534 | |||
| AK159628.1 | - | 3 | 510 | |||
| AK159792.1 | - | 3 | 510 | |||
| AK159962.1 | - | 3 | 513 | |||
| AK160101.1 | - | 3 | 510 | |||
| AK160725.1 | - | 3 | 541 | |||
| AK194069.1 | - | 87 | ||||
| BC004731.1 | - | 3 | 493 | 3583154 | ||
| BC010320.1 | - | 3 | 525 | 3599190 | ||
| BC021786.1 | - | 3 | 531 | 4458918 | ||
| U76253.1 | - | 3 | 522 | |||

Itm2b
Genomic Map