siRNA Details
| Gene Symbol: Itpr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010585.5 | NP_034715.3 | 54 | 7353 | ||
| GenBank mRNA | AK044776.1 | - | 2 | 146 | ||
| BC003271.1 | - | 6 | 712 | |||
| BC068269.1 | - | 4 | 449 | |||
| BQ180028.1 | - | 4 | 463 | |||
| CN692248.1 | - | 3 | 406 | |||
| M21530.1 | - | 3 | 256 | |||
| X15373.1 | - | 54 | 7331 | |||

Itpr1
Genomic Map