siRNA Details
Gene Symbol: Kcnab2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252654.1 | NP_001239583.1 | 6 | 459 | ||
NM_001252655.1 | NP_001239584.1 | 6 | 488 | |||
NM_001252656.1 | NP_001239585.1 | 7 | 530 | |||
NM_010598.3 | NP_034728.2 | 7 | 501 | |||
GenBank mRNA | AK143166.1 | - | 6 | 436 | ||
AK152613.1 | - | 7 | 453 | |||
AK157053.1 | - | 6 | 452 | |||
BC039178.1 | - | 7 | 431 | 4503373 | ||
L48983.1 | - | 7 | 493 | |||
U65592.1 | - | 6 | 296 |
