siRNA Details
| Gene Symbol: Kcnk5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021542.4 | NP_067517.1 | 1 | 453 | ||
| GenBank mRNA | AF259395.1 | - | 1 | 123 | ||
| AF319542.1 | - | 1 | 192 | |||
| AK085330.1 | - | 1 | 454 | |||
| AK132172.1 | - | 1 | 481 | |||
| AK165211.1 | - | 1 | 452 | |||
| AK172608.1 | - | 1 | 454 | |||
| BC034012.1 | - | 1 | 455 | 4239113 | ||
| BC051088.1 | - | 1 | 422 | |||
| BC058164.1 | - | 1 | 444 | 5323480 | ||

Kcnk5
Genomic Map