siRNA Details
| Gene Symbol: Kcnn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163510.1 | NP_001156982.1 | 5 | 1073 | ||
| NM_008433.4 | NP_032459.3 | 5 | 1073 | |||
| GenBank mRNA | AB128930.1 | - | 3 | 836 | ||
| AF042487.1 | - | 5 | 939 | |||
| AF072884.1 | - | 5 | 899 | |||
| AK010943.1 | - | 5 | 1074 | |||
| AK085752.1 | - | 5 | 890 | |||
| AK155910.1 | - | 5 | 1038 | |||
| BC010274.1 | - | 5 | 1022 | 3589411 | ||

Kcnn4
Genomic Map