siRNA Details
| Gene Symbol: Kcnn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163510.1 | NP_001156982.1 | 4 | 891 | ||
| NM_008433.4 | NP_032459.3 | 4 | 891 | |||
| GenBank mRNA | AB128930.1 | - | 2 | 654 | ||
| AF042487.1 | - | 4 | 757 | |||
| AF072884.1 | - | 4 | 717 | |||
| AK010943.1 | - | 4 | 892 | |||
| AK085752.1 | - | 4 | 708 | |||
| AK155910.1 | - | 4 | 856 | |||
| BC010274.1 | - | 4 | 840 | 3589411 | ||

Kcnn4
Genomic Map