siRNA Details
| Gene Symbol: Kdr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010612.2 | NP_034742.2 | 17 | 2650 | ||
| GenBank mRNA | BC020530.1 | - | 17 | 2650 | 4238984 | |
| S53103.1 | - | 17 | 2661 | |||
| X59397.1 | - | 17 | 2597 | |||
| X70842.1 | - | 17 | 2675 | |||

Kdr
Genomic Map