siRNA Details
| Gene Symbol: Kera | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008438.3 | NP_032464.1 | 2 | 610 | ||
| GenBank mRNA | AF022256.1 | - | 2 | 614 | ||
| AK086197.1 | - | 2 | 634 | |||
| BC114980.1 | - | 2 | 476 | 40046884 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kera| Gene Symbol: Kera | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008438.3 | NP_032464.1 | 2 | 610 | ||
| GenBank mRNA | AF022256.1 | - | 2 | 614 | ||
| AK086197.1 | - | 2 | 634 | |||
| BC114980.1 | - | 2 | 476 | 40046884 | ||