siRNA Details
| Gene Symbol: Kif7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291222.1 | NP_001278151.1 | 17 | 3633 | ||
| NM_010626.3 | NP_034756.2 | 17 | 3630 | |||
| GenBank mRNA | BC034093.1 | - | 7 | 1205 | ||
| BC141045.1 | - | 17 | 3631 | 9055657 | ||
| BC145222.1 | - | 17 | 3634 | 9053773 | ||

Kif7
Genomic Map