siRNA Details
| Gene Symbol: Kifc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010630.2 | NP_034760.2 | 5 | 620 | ||
| GenBank mRNA | AK144304.1 | - | 6 | 779 | ||
| D49545.1 | - | 5 | 696 | |||
| U92949.1 | - | 5 | 610 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kifc2| Gene Symbol: Kifc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010630.2 | NP_034760.2 | 5 | 620 | ||
| GenBank mRNA | AK144304.1 | - | 6 | 779 | ||
| D49545.1 | - | 5 | 696 | |||
| U92949.1 | - | 5 | 610 | |||