siRNA Details
| Gene Symbol: Uhmk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010633.3 | NP_034763.3 | 2 | 581 | ||
| GenBank mRNA | AK013347.1 | - | 2 | 586 | ||
| AK030152.1 | - | 398 | ||||
| BC058732.1 | - | 2 | 581 | 6414877 | ||
| X82320.1 | - | 1 | 67 | |||
| Y10725.2 | - | 2 | 438 | |||

Uhmk1
Genomic Map