siRNA Details
| Gene Symbol: Klk1b16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008454.2 | NP_032480.1 | 4 | 1064 | ||
| GenBank mRNA | BC012237.1 | - | 4 | 535 | 4165443 | |
| J03877.1 | - | 4 | 1065 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Klk1b16| Gene Symbol: Klk1b16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008454.2 | NP_032480.1 | 4 | 1064 | ||
| GenBank mRNA | BC012237.1 | - | 4 | 535 | 4165443 | |
| J03877.1 | - | 4 | 1065 | |||