siRNA Details
| Gene Symbol: Klrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010654.3 | NP_034784.1 | 6 | 546 | ||
| GenBank mRNA | AF030311.1 | - | 5 | 526 | ||
| AF030312.1 | - | 6 | 575 | |||
| AF039025.1 | - | 6 | 540 | |||
| AF057714.1 | - | 5 | 526 | |||
| AK136548.1 | - | 6 | 547 | |||
| AK162976.1 | - | 7 | 607 | |||
| BC117112.1 | - | 6 | 564 | 40126431 | ||
| BC120854.1 | - | 6 | 566 | 40129777 | ||

Klrd1
Genomic Map