siRNA Details
| Gene Symbol: Sspn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010656.2 | NP_034786.1 | 1, 2 | 215 | ||
| GenBank mRNA | AK148179.1 | - | 1, 2 | 217 | ||
| AK162864.1 | - | 1, 2 | 240 | |||
| AK163041.1 | - | 1, 2 | 241 | |||
| AK164430.1 | - | 1, 2 | 241 | |||
| AK164529.1 | - | 1, 2 | 187 | |||
| BC021484.1 | - | 1, 2 | 240 | 5344514 | ||
| U02487.1 | - | 1, 2 | 215 | |||

Sspn
Genomic Map