siRNA Details
Gene Symbol: Lama3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010680.1 | NP_034810.1 | 38, 39 | 4992 | ||
GenBank mRNA | X84014.1 | - | 19, 20 | 2710 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Lama3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010680.1 | NP_034810.1 | 38, 39 | 4992 | ||
GenBank mRNA | X84014.1 | - | 19, 20 | 2710 |