siRNA Details
| Gene Symbol: Lamb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008483.3 | NP_032509.2 | 17 | 2196 | ||
| GenBank mRNA | AK140351.1 | - | 17 | 2175 | ||
| AK158444.1 | - | 17 | 2169 | |||
| BC026051.1 | - | 17 | 2123 | 4036357 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lamb2| Gene Symbol: Lamb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008483.3 | NP_032509.2 | 17 | 2196 | ||
| GenBank mRNA | AK140351.1 | - | 17 | 2175 | ||
| AK158444.1 | - | 17 | 2169 | |||
| BC026051.1 | - | 17 | 2123 | 4036357 | ||