siRNA Details
| Gene Symbol: Lamb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277928.1 | NP_001264857.1 | 5, 6 | 524 | ||
| NM_008484.2 | NP_032510.2 | 5, 6 | 563 | |||
| GenBank mRNA | AK155769.1 | - | 1376 | |||
| BC008516.1 | - | 5, 6 | 524 | 3592888 | ||
| BY248755.1 | - | 2, 3 | 113 | |||
| U43298.1 | - | 5, 6 | 539 | |||

Lamb3
Genomic Map