siRNA Details
| Gene Symbol: Lbx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010691.5 | NP_034821.2 | 2 | 432 | ||
| GenBank mRNA | AB108497.1 | - | 2 | 432 | ||
| AK144562.1 | - | 2 | 277 | |||
| BC119177.1 | - | 2 | 521 | 8733926 | ||
| BC120586.1 | - | 2 | 521 | 40129509 | ||
| X90829.1 | - | 1 | 1565 | |||

Lbx1
Genomic Map