siRNA Details
| Gene Symbol: Lbx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010692.3 | NP_034822.1 | 1 | 92 | ||
| GenBank mRNA | AF146150.1 | - | 1 | 33 | ||
| AK002897.1 | - | 1 | 92 | |||
| BC109347.2 | - | 1 | 87 | 40055655 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lbx2| Gene Symbol: Lbx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010692.3 | NP_034822.1 | 1 | 92 | ||
| GenBank mRNA | AF146150.1 | - | 1 | 33 | ||
| AK002897.1 | - | 1 | 92 | |||
| BC109347.2 | - | 1 | 87 | 40055655 | ||