siRNA Details
| Gene Symbol: Lrrn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010732.4 | NP_034862.1 | 2 | 805 | ||
| GenBank mRNA | AK036271.1 | - | 2 | 807 | ||
| AK039216.1 | - | 2 | 806 | |||
| AY362823.1 | - | 1 | 394 | |||
| BC056458.1 | - | 2 | 547 | 5685881 | ||

Lrrn2
Genomic Map