siRNA Details
| Gene Symbol: Lst1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010734.2 | NP_034864.2 | 4 | 297 | ||
| GenBank mRNA | AF000428.1 | - | 4 | 248 | ||
| AI182980.1 | - | 4 | 296 | |||
| AI323110.1 | - | 3 | 202 | |||
| AI482475.1 | - | 1 | 118 | |||
| AK146409.1 | - | 3 | 155 | |||
| BC086881.1 | - | 4 | 291 | 6747449 | ||
| M18187.1 | - | 4 | 289 | |||
| U72643.1 | - | 4 | 285 | |||
| U72644.1 | - | 4 | 285 | |||

Lst1
Genomic Map