siRNA Details
| Gene Symbol: Ltbp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008520.2 | NP_032546.2 | 7 | 2083 | ||
| GenBank mRNA | AK038587.1 | - | 6 | 815 | ||
| AK080869.1 | - | 7 | 2083 | |||
| AK131126.1 | - | 3 | 2622 | |||
| AK178255.1 | - | 2 | 104 | |||
| BC127051.1 | - | 6 | 682 | |||

Ltbp3
Genomic Map