siRNA Details
| Gene Symbol: Ltbp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008520.2 | NP_032546.2 | 26 | 4423 | ||
| GenBank mRNA | AK007774.1 | - | 6 | 709 | ||
| BC040761.1 | - | 7 | 1331 | |||
| BC054420.1 | - | 7 | 1313 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ltbp3| Gene Symbol: Ltbp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008520.2 | NP_032546.2 | 26 | 4423 | ||
| GenBank mRNA | AK007774.1 | - | 6 | 709 | ||
| BC040761.1 | - | 7 | 1331 | |||
| BC054420.1 | - | 7 | 1313 | |||